Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,6-Diethylaniline 98%

Irritant
2,6-Diethylaniline 98%
Certificates
2,6-Diethylaniline 98%

2 Options Available Below

Product Details & Documents
  • CAS Number:579-66-8
  • Formula:C₁₀H₁₅N
  • MDL Number:MFCD00007753
  • Molecular Weight:149.24 g/mol
  • Boiling Pt:243 °C (1013 hPa)
  • Melting Pt:3…4 °C
  • Density:0.9 g/cm³
  • Flash Pt:123 °C
  • Storage Temperature:Ambient
  • EINECS:209-445-7

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear yellow to red-brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5446 to 1.5466 (20°C, 589 nm)
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 55,40 €
    55,40 €
    1 * 250 mL
       55,40 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 141,00 €
    141,00 €
    1 * 1 L
       141,00 €