


2,4-Xylidine 99%
:95-68-1
:
:C₈H₁₁N
:MFCD00007738
:121.18 g/mol
:12,10217
:218 °C (1013 hPa)
:16 °C
:0.98 g/cm³
:90 °C
:Ambient
:202-440-0
:1711
:6.1,II
:GHS06
:GHS08
:GHS09
- Appearance:Clear yellow to red-brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5580 to 1.5600 (20°C, 589 nm)
- GC:≥98.5 %
- Titration with HClO4:98.5 to 101.5 %