

α,α,α-Trifluoro-p-xylene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6140-17-6
:
Formula:C₈H₇F₃
MDL Number:MFCD00075476
:160.14 g/mol
Boiling Pt:129 °C (760 mmHg)
Density:1.144 g/cm³
Flash Pt:27 °C
Storage Temperature:Ambient
EINECS:228-124-2
UN:1993
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4250 to 1.4270 (20°C, 589 nm)
- GC:≥97.5 %