α,α'-Dichloro-p-xylene
Catalog # MOLE38947698-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:175.06 g/mol
Boiling Pt:254 °C (1013 hPa)
Melting Pt:100 °C
Density:1.432 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:210-782-7
UN:2928
ADR:6.1,II
CAS Number:623-25-6
Formula:C₈H₈Cl₂
MDL Number:MFCD00000920