1,3-Dicyclohexylurea, Sigma-Aldrich®
SIALD80800-25G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
: N,N′-Dicyclohexylurea, N,N'-Dicyclohexylurea
:
:M-Clarity™ quality level = MQ100
:224.35 g/mol
:219-213-7
:2387-23-7
:C₁₃H₂₄N₂O
:MFCD00003829