1,3-Diethylurea, Sigma-Aldrich®
Catalog # SIALD101087-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:116.16 g/mol
:M-Clarity™ quality level = MQ100
EINECS:210-811-3
CAS Number:623-76-7
Formula:C₅H₁₂N₂O
MDL Number:MFCD00009028