Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-(-)-4-Methoxy-α-methylbenzylamine
(S)-(-)-4-Methoxy-α-methylbenzylamine, Sigma-Aldrich®
(S)-(-)-4-Methoxy-α-methylbenzylamine, Sigma-Aldrich®
(S)-(-)-4-Methoxy-α-methylbenzylamine, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
(S)-(-)-4-Methoxy-α-methylbenzylamine, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
41851-59-6
:
Formula:
C₉H₁₃NO
MDL Number:
MFCD00671660
:
151.21 g/mol
UN:
2735
ADR:
8,III