3,4-Dichlorophenethylamine, Sigma-Aldrich®
Catalog # SIAL560421-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:190.07 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:21581-45-3
Formula:C₈H₉Cl₂N
MDL Number:MFCD00060617