2,6-Dichlorophenethylamine, Sigma-Aldrich®
Glass bottle for solids, 5 g
Catalog # SIAL560413-5G
Supplier: Merck
2,6-Dichlorophenethylamine, Sigma-Aldrich®
Catalog # SIAL560413-5G
Supplier: Merck
CAS Number: 14573-23-0
Jump to:
Product Details & Documents
Synonyms: 2-(2-Aminoethyl)-1,3-dichlorobenzene, 1-Amino-2-(2,6-dichlorophenyl)ethane, 2-(2,6-Dichlorophenyl)ethylamine, 2,6-Dichlorophenethylamine
- CAS Number:14573-23-0
- Formula:C₈H₉Cl₂N
- MDL Number:MFCD00060612
- Molecular Weight:190.07 g/mol
- :M-Clarity™ quality level = MQ100
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



