3,3-Diphenylpropylamine, Sigma-Aldrich®
Catalog # SIAL136298-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:211.31 g/mol
:M-Clarity™ quality level = MQ100
EINECS:226-984-3
CAS Number:5586-73-2
Formula:C₁₅H₁₇N
MDL Number:MFCD00008202