
2,6-Dimethylbenzoic acid 99%
Catalog # ACRO169010010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- Melting point:114 to 117 °C
- Titration with NaOH:≥98.5 %
Specification Test Results
About this item
:
:150.18 g/mol
Boiling Pt:155 - 160 °C (17 -rr)
Melting Pt:114…117 °C
EINECS:211-177-0
:GHS07
Storage Temperature:Ambient
CAS Number:632-46-2
Formula:C₉H₁₀O₂
MDL Number:MFCD00002483