3,3-Diphenylpropylamine
Glass bottle for solids, 25 g
Catalog # MOLE23812498-25G
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 3,3-Diphenylpropan-1-amine
- CAS Number:5586-73-2
- Formula:C₁₅H₁₇N
- MDL Number:MFCD00008202
- Molecular Weight:211.31 g/mol
- EINECS:226-984-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



