Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dimethoxyphenethylamine
3,4-Dimethoxyphenethylamine
3,4-Dimethoxyphenethylamine
3,4-Dimethoxyphenethylamine
Catalog # MOLEM66970771
Supplier:
Molekula
CAS Number:
Expand image
3,4-Dimethoxyphenethylamine
Catalog # MOLEM66970771
Supplier:
Molekula
CAS Number:
Specifications
Pk:
100 g
Seq:
3401
Specifications
About this item
Product Details
:
:
181.23
EINECS:
204-376-9
CAS Number:
120-20-7
Formula:
C₁₀H₁₅NO₂
MDL Number:
MFCD00008188
Product Details