(1S,2R)-(+)-2-Amino-1,2-diphenylethanol
Catalog # MOLE72095716-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:213.28 g/mol
CAS Number:23364-44-5
Formula:C₁₄H₁₅NO
MDL Number:MFCD00074959