Order Entry
Export
ContactUsLinkComponent
4-Bromothiophene-2-carbaldehyde
4-Bromothiophene-2-carbaldehyde
Catalog # MOLEM23521866
Supplier:  Molekula
CAS Number:  
4-Bromothiophene-2-carbaldehyde
Catalog # MOLEM23521866
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :191.05 g/mol
EINECS:242-577-3
CAS Number:18791-75-8
Formula:C₅H₃BrOS
MDL Number:MFCD00005431