Order Entry
Export
ContactUsLinkComponent
2,4-Dimethylbenzoic acid 98%
Irritant
2,4-Dimethylbenzoic acid 98%
Certificates
undefined
Irritant
2,4-Dimethylbenzoic acid 98%

Specification Test Results

  • Appearance:
    White to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    122 to 127 °C
  • Titration with NaOH:
    ≥97.5 %

Specification Test Results

Related Information