Order Entry
Export
Orders LinkContactUsLinkComponent
Lidocaine

Lidocaine

Glass bottle for solids, 1 mg
Catalog # FITZ10-L40-M24072
Lidocaine
Catalog # FITZ10-L40-M24072
CAS Number:  137-58-6
Lidocaine

Glass bottle for solids, 1 mg

CAS Number:  137-58-6
Price
Inquire For Price
Log in for availability
1 * 1 mg
Product Details & Documents
  • CAS Number:137-58-6
  • Formula:C₁₄H₂₂N₂O
  • MDL Number:MFCD00026733
  • Molecular Weight:234.34 g/mol
  • EINECS:205-302-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 mg

Specifications