Order Entry
Export
ContactUsLinkComponent
1,3-Diphenylurea
1,3-Diphenylurea
Catalog # EHERC12921500
Supplier:  EHRENSTORFER
CAS Number:  
1,3-Diphenylurea
Catalog # EHERC12921500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :212.25 g/mol
EINECS:203-003-7
CAS Number:102-07-8
Formula:C₁₃H₁₂N₂O
MDL Number:MFCD00003017