
Cyclohexanone oxime
Supplier: CHEMPUR
Certificates
About this item
CAS Number:100-64-1
:
Formula:C₆H₁₁NO
MDL Number:MFCD00001660
:113.16 g/mol
Boiling Pt:206 °C (1013 hPa)
Melting Pt:88…90 °C
Density:1,13 g/cm³ (20 °C)
EINECS:202-874-0
:GHS07