Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acetophenone oxime
Acetophenone oxime
Acetophenone oxime
Acetophenone oxime
Supplier:
CHEMPUR
Certificates
Expand image
Acetophenone oxime
Supplier:
CHEMPUR
About this item
CAS Number:
613-91-2
:
Formula:
C₈H₉NO
MDL Number:
MFCD00013931
:
135.17 g/mol
EINECS:
210-360-2