Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Diethylurea ≥97%

1,3-Diethylurea ≥97%
Certificates
1,3-Diethylurea ≥97%
1,3-Diethylurea ≥97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 623-76-7 Molecular Formula: C5H12N2O Formula Weight: 116.16 Melting Point: 109-113° Beilstein Reference: 1744741

Synonyms:  N,N'-Diethylurea
  • CAS Number:623-76-7
  • Formula:C₅H₁₂N₂O
  • MDL Number:MFCD00009028
  • Molecular Weight:116.16 g/mol
  • Melting Pt:109…113 °C
  • Storage Temperature:Ambient
  • EINECS:210-811-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 52,90 €
    52,90 €
    1 * 5 g
       52,90 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 152,00 €
    152,00 €
    1 * 25 g
       152,00 €