Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,6-Dimethylbenzonitrile

Supplier:   Thermo Scientific
2,6-Dimethylbenzonitrile
Supplier:  Thermo Scientific
Certificates
2,6-Dimethylbenzonitrile

3 Options Available Below

Product Details & Documents
  • CAS Number:6575-13-9
  • Formula:C₉H₉N
  • MDL Number:MFCD00037166
  • Molecular Weight:131.18 g/mol
  • EINECS:229-503-5
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g