Order Entry
Export
Orders LinkContactUsLinkComponent
3,4-Dichlorophenethylamine 99%

3,4-Dichlorophenethylamine 99%

Irritant
3,4-Dichlorophenethylamine 99%
Certificates
3,4-Dichlorophenethylamine 99%

2 Options Available Below

Product Details & Documents
  • CAS Number:21581-45-3
  • Formula:C₈H₉Cl₂N
  • MDL Number:MFCD00060617
  • Molecular Weight:190.07 g/mol
  • Boiling Pt:280 °C
  • Density:1.268 g/cm³
  • Flash Pt:110 °C
  • Storage Temperature:Ambient
  • ADR:8,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5655 to 1.5675 (20°C, 589 nm)
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 136,00 €
    136,00 €
    1 * 5 g
       136,00 €

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 455,00 €
    455,00 €
    1 * 25 g
       455,00 €