Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dimethylbenzaldehyde ≥98%

3,5-Dimethylbenzaldehyde ≥98%

3,5-Dimethylbenzaldehyde ≥98%
Certificates
3,5-Dimethylbenzaldehyde ≥98%
3,5-Dimethylbenzaldehyde ≥98%

2 Options Available Below

Product Details & Documents

Refractive Index: 1.5375
Melting Point: 8-10°
Boiling Point: 220-222°
Beilstein Reference: 2038840
MDL No.: MFCD00082776

  • CAS Number:5779-95-3
  • Formula:C₉H₁₀O
  • MDL Number:MFCD00082777
  • Molecular Weight:134.18 g/mol
  • Boiling Pt:220…222 °C
  • Melting Pt:8…10 °C
  • Density:1.09 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:675-424-8
Product Family Options

Product Information