Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dimethylbenzaldehyde
3,4-Dimethylbenzaldehyde
3,4-Dimethylbenzaldehyde
3,4-Dimethylbenzaldehyde
Supplier:
Thermo Scientific
Certificates
Expand image
3,4-Dimethylbenzaldehyde
Supplier:
Thermo Scientific
About this item
CAS Number:
5973-71-7
:
Formula:
C₉H₁₀O
MDL Number:
MFCD00016612
:
134.18 g/mol
Boiling Pt:
68 °C (0.8 mmHg)
Density:
1.012 g/cm³ (20 °C)
EINECS:
227-770-2