α'-Bromo-α,α,α-trifluoro-p-xylene 98%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 4-(Bromomethyl)benzotrifluoride, 1-(Bromomethyl)-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)benzyl bromide
- CAS Number:402-49-3
- Formula:C₈H₆BrF₃
- MDL Number:MFCD00000403
- Molecular Weight:239.03 g/mol
- Boiling Pt:65…69 °C (5 mmHg)
- Melting Pt:30…34 °C
- Density:1.54 g/cm³
- Flash Pt:88 °C
- Storage Temperature:Ambient
- EINECS:206-947-8
- ADR:8,III
- :Density: 1.54 g/cm³ (20 °C)
- UN:1760
3 Options Available Below
Product Details & Documents
Synonyms: 4-(Bromomethyl)benzotrifluoride, 1-(Bromomethyl)-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)benzyl bromide
- CAS Number:402-49-3
- Formula:C₈H₆BrF₃
- MDL Number:MFCD00000403
- Molecular Weight:239.03 g/mol
- Boiling Pt:65…69 °C (5 mmHg)
- Melting Pt:30…34 °C
- Density:1.54 g/cm³
- Flash Pt:88 °C
- Storage Temperature:Ambient
- EINECS:206-947-8
- ADR:8,III
- :Density: 1.54 g/cm³ (20 °C)
- UN:1760
Specifications for Product Family
Specification Test Results
- Appearance:White to light yellow Low melting crystalline solid
- Infrared spectrum:Conforms
- Melting point:30 to 34 °C
- GC:≥97.5 %



