
4-Bromo-o-xylene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:583-71-1
:
Formula:C₈H₉Br
MDL Number:MFCD00000071
:185.06 g/mol
Boiling Pt:214…215 °C
Density:1.370
Flash Pt:78 °C (172 °F)
Storage Temperature:Ambient
EINECS:209-518-3
UN:1701
ADR:6.1,II
:GHS07