
4-Bromo-m-xylene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:583-70-0
:
Formula:C₈H₉Br
MDL Number:MFCD00000071
:185.06 g/mol
Boiling Pt:212…213 °C
Melting Pt:–1 °C
Density:1.366
Flash Pt:78 °C (174 °F)
EINECS:209-517-8
:GHS07
Storage Temperature:Ambient