
2-Bromo-m-xylene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:576-22-7
:
Formula:C₈H₉Br
MDL Number:MFCD00000075
:185.06 g/mol
Boiling Pt:206 °C (760 mmHg)
Melting Pt:–10 °C
Density:1.38 g/cm³
Flash Pt:73 °C
EINECS:209-397-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5540 to 1.5560 (20°C, 589 nm)
- GC:≥97.5 %
- Stabilizer:Approx. 100 ppm BHT