
3-Bromo-o-xylene 99%
ACRO107580050
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5590 to 1.5610 (20°C, 589 nm)
- GC:≥98.5 %
:
:185.06 g/mol
:214 °C (1013 hPa)
:1.36 g/cm³
:80 °C
:209-398-2
:6.1,II
:GHS07
:576-23-8
:C₈H₉Br
:MFCD00000069