

α-Bromo-p-xylene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:104-81-4
:
Formula:C₈H₉Br
MDL Number:MFCD00000181
Merck Index:14,10090
Boiling Pt:218…220 °C
Melting Pt:33…36 °C
Density:1.324
Flash Pt:97 °C (206 °F)
UN:3417
ADR:6.1,II
:185.06 g/mol
EINECS:203-240-6
:GHS06
:GHS05
Storage Temperature:Refrigerator