

α-Bromo-p-xylene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:104-81-4
:
Formula:C₈H₉Br
MDL Number:MFCD00000181
Merck Index:12,10223
Boiling Pt:218…220 °C (760 mmHg)
Melting Pt:33…37 °C
Density:1,324 g/cm³ (20 °C)
Flash Pt:97 °C
UN:3417
ADR:6.1,II
:185.06 g/mol
EINECS:203-240-6
:GHS07
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Crystalline mass
- Infrared spectrum:Conforms
- Melting point:33 to 37 °C
- GC:≥97.5 %