Order Entry
Export
ContactUsLinkComponent
2,5-Dibromo-p-xylene 98+%
Irritant
2,5-Dibromo-p-xylene 98+%
Certificates
undefined
Irritant
2,5-Dibromo-p-xylene 98+%

Specification Test Results

  • Appearance:
    White to beige to light orange Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    72 to 76 °C
  • GC:
    ≥98.0 %

Specification Test Results

Related Information