

α,α'-Dibromo-o-xylene 96%
:91-13-4
:
:C₈H₈Br₂
:MFCD00000175
:263.96 g/mol
:140 °C (20 mmHg)
:91…96 °C
:1.96 g/cm³
:Ambient
:202-042-7
:2928
:6.1,II
:GHS07
:GHS05
- Appearance:White to brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:91 to 96 °C
- GC:≥95 %