1,3-Diphenylacetone ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 102-04-5 Molecular Formula: C15H14O Formula Weight: 210.28 Flash Point: 110°(230°F) Melting Point: 32-36° Boiling Point: 330° Beilstein Reference: 974767 MDL No.: MFCD00004795
Synonyms: 1,3-Diphenylpropan-2-one, 1,3-Diphenyl-2-propanone
- CAS Number:102-04-5
- Formula:(C₆H₅)CH₂C(O)CH₂(C₆H₅)
- MDL Number:MFCD00004795
- Molecular Weight:210.28 g/mol
- Boiling Pt:330 °C
- Melting Pt:32…36 °C
- Density:1.04
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:203-000-0
3 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 102-04-5 Molecular Formula: C15H14O Formula Weight: 210.28 Flash Point: 110°(230°F) Melting Point: 32-36° Boiling Point: 330° Beilstein Reference: 974767 MDL No.: MFCD00004795
Synonyms: 1,3-Diphenylpropan-2-one, 1,3-Diphenyl-2-propanone
- CAS Number:102-04-5
- Formula:(C₆H₅)CH₂C(O)CH₂(C₆H₅)
- MDL Number:MFCD00004795
- Molecular Weight:210.28 g/mol
- Boiling Pt:330 °C
- Melting Pt:32…36 °C
- Density:1.04
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:203-000-0



