


Toluene 99+%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-88-3
:
Formula:C₆H₅CH₃
MDL Number:MFCD00008512
:92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.866 g/cm³
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
:GHS07
:GHS08
:GHS02
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.05 % (K.F.)
- GC:≥99.0 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.003 %
- UV:at 280 nm A: ≤0.1
- pH:6 to 9 (0.4 mol sol)
- Titration with HClO4:98.5 to 102 %
- Heavy metals (as Pb):≤10 ppm