Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dinitrotoluene
2,6-Dinitrotoluene, Sigma-Aldrich®
2,6-Dinitrotoluene, Sigma-Aldrich®
:
Merck
2,6-Dinitrotoluene, Sigma-Aldrich®
:
Merck
:
606-20-2
:
:
C₇H₆N₂O₄
:
MFCD00007158
:
182.14 g/mol
:
300 °C (1013 hPa)
:
65.2 °C
:
1.538 g/cm³ (15 °C)
:
Ambient
:
210-106-0
:
6.1,II
:
M-Clarity™ quality level = MQ200
:
3454
:
GHS06
:
GHS08