Order Entry
Export
ContactUsLinkComponent
3,4-Dinitrotoluene
Toxic
Health
Environment
3,4-Dinitrotoluene
Catalog # EHERC12786500
Supplier:  EHRENSTORFER
CAS Number:  
Toxic
Health
Environment
3,4-Dinitrotoluene
Catalog # EHERC12786500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
Boiling Pt:351 °C (1013 hPa)
Melting Pt:61 °C
Density:1.2594 g/cm³ (111 °C)
ADR:6.1,II
 :182.14 g/mol
EINECS:210-222-1
UN:3454
 :GHS06
 :GHS08
 :GHS09
CAS Number:610-39-9
Formula:C₇H₆N₂O₄