
α,α,α-Trifluoro-3,5-dinitrotoluene
Catalog # MOLE13027435-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:236.11 g/mol
Boiling Pt:127…129 °C (9 mmHg)
Melting Pt:50 °C
EINECS:206-935-2
:GHS07
Storage Temperature:Ambient
CAS Number:401-99-0
Formula:C₇H₃F₃N₂O₄
MDL Number:MFCD00007233