Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
α-Chloro-3-nitrotoluene
α-Chloro-3-nitrotoluene
α-Chloro-3-nitrotoluene
Corrosive
α-Chloro-3-nitrotoluene
Catalog # MOLEM37384152
Supplier:
Molekula
CAS Number:
Expand image
Corrosive
α-Chloro-3-nitrotoluene
Catalog # MOLEM37384152
Supplier:
Molekula
CAS Number:
About this item
Product Details
Product Details