Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
α,3,4-Trichlorotoluene
α,3,4-Trichlorotoluene ≥98%
α,3,4-Trichlorotoluene ≥98%
:
Thermo Fisher Scientific
α,3,4-Trichlorotoluene ≥98%
:
Thermo Fisher Scientific
:
102-47-6
:
:
C₇H₅Cl₃
:
MFCD00000906
:
195.47 g/mol
:
99…101 °C (2 mmHg)
:
–3 °C
:
1.416
:
155 °C (311 °F)
:
Ambient
:
203-033-0
:
3265
:
8,II
:
GHS05
:
GHS07
msds_ALFAA17672
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years