

α,3,4-Trichlorotoluene 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:102-47-6
:
Formula:C₇H₅Cl₃
MDL Number:MFCD00000906
:195.47 g/mol
Boiling Pt:122…124 °C (14 mmHg)
Melting Pt:–3 °C
Density:1.41 g/cm³
Flash Pt:155 °C
Storage Temperature:Ambient
EINECS:203-033-0
UN:3265
ADR:8,III
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5756 to 1.5776 (20°C, 589 nm)
- GC:≥98.0 %