α,2,4-Trichlorotoluene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:94-99-5
:
Formula:C₇H₅Cl₃
MDL Number:MFCD00000895
:195.47 g/mol
Boiling Pt:248 °C (1013 hPa)
Melting Pt:–2.7 °C
Density:1.4 g/cm³
Flash Pt:190 °C
Storage Temperature:Ambient
EINECS:202-381-0
UN:3265
ADR:8,III
:GHS07
:GHS05
:GHS09
Specification Test Results
- Appearance:Clear colorless to light yellow Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.5750 to 1.5770 (20°C, 589 nm)
- GC:≥98.5 %
- Enantiomeric excess:≥98.0 %
- Specific optical rotation:+7° to +9° (20°C, 589 nm) (c=1, chloroform)