Order Entry
Export
ContactUsLinkComponent
2,6-Dichlorotoluene
Environment
2,6-Dichlorotoluene
Catalog # EHERC12505900
Supplier:  EHRENSTORFER
CAS Number:  
Environment
2,6-Dichlorotoluene
Catalog # EHERC12505900
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :161.03 g/mol
Boiling Pt:200 °C (1013 hPa)
Melting Pt:2 °C
Density:1.2686 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-269-7
UN:3082
ADR:9,III
 :GHS09
CAS Number:118-69-4
Formula:C₇H₆Cl₂
MDL Number:MFCD00000576