
2,6-Dichlorotoluene ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:118-69-4
:
Formula:C₇H₆Cl₂
MDL Number:MFCD00000576
:161.03 g/mol
Boiling Pt:196…203 °C
Melting Pt:2 °C
Density:1.254
Flash Pt:82 °C (179 °F)
Storage Temperature:Ambient
EINECS:204-269-7
UN:3082
ADR:9,III
:GHS09