Order Entry
Export
ContactUsLinkComponent
2,6-Dichlorotoluene 99+%
Irritant
Environment
2,6-Dichlorotoluene 99+%
Certificates
undefined
Irritant
Environment
2,6-Dichlorotoluene 99+%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5500 to 1.5520 (20°C, 589 nm)
  • GC:
    ≥99.0 %

Specification Test Results

Related Information