Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Nitro-o-toluic acid
3-Nitro-o-toluic acid
3-Nitro-o-toluic acid
3-Nitro-o-toluic acid
Supplier:
Thermo Scientific
Certificates
Expand image
3-Nitro-o-toluic acid
Supplier:
Thermo Scientific
About this item
CAS Number:
1975-50-4
:
Formula:
C₈H₇NO₄
MDL Number:
MFCD00007160
:
181.15 g/mol
Melting Pt:
181…184 °C
Storage Temperature:
Ambient
EINECS:
217-826-4