Order Entry
Export
ContactUsLinkComponent
α-Bromo-2,6-difluorotoluene 97%
Corrosive
α-Bromo-2,6-difluorotoluene 97%
Certificates
undefined
Corrosive
α-Bromo-2,6-difluorotoluene 97%

Specification Test Results

  • Appearance:
    White to light beige Crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    51 to 55 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information