Order Entry
Export
Orders LinkContactUsLinkComponent
2-Bromotoluene

2-Bromotoluene

Glass bottle for solids, 100 g
Catalog # MOLE52688778-100G
Supplier:   Molekula
2-Bromotoluene
Catalog # MOLE52688778-100G
Supplier:  Molekula
CAS Number:  95-46-5
2-Bromotoluene

Glass bottle for solids, 100 g

CAS Number:  95-46-5
Price
130,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:95-46-5
  • Formula:C₇H₇Br
  • MDL Number:MFCD00000068
  • Molecular Weight:171.04 g/mol
  • Merck Index:13,01424
  • Boiling Pt:181.5 °C (1013 hPa)
  • Melting Pt:–27 °C
  • Density:1.4192 g/cm³ (20 °C)
  • Flash Pt:79 °C
  • Storage Temperature:Refrigerator
  • EINECS:202-421-7
  • UN:3082
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications