2-Bromotoluene ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 95-46-5 Molecular Formula: C7H7Br Formula Weight: 171.04 Flash Point: 78°(172°F) Refractive Index: 1.5560 Melting Point: -27° Boiling Point: 179-181° Beilstein Reference: 1904176 Merck Reference: 14,1439 MDL No.: MFCD00000068
- CAS Number:95-46-5
- Formula:C₇H₇Br
- MDL Number:MFCD00000068
- Molecular Weight:171.04 g/mol
- Merck Index:14,1439
- Boiling Pt:179…181 °C
- Melting Pt:–27 °C
- Density:1.420
- Flash Pt:78 °C (172 °F)
- Storage Temperature:Ambient
- EINECS:202-421-7
- UN:3082
- ADR:9,III
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 95-46-5 Molecular Formula: C7H7Br Formula Weight: 171.04 Flash Point: 78°(172°F) Refractive Index: 1.5560 Melting Point: -27° Boiling Point: 179-181° Beilstein Reference: 1904176 Merck Reference: 14,1439 MDL No.: MFCD00000068
- CAS Number:95-46-5
- Formula:C₇H₇Br
- MDL Number:MFCD00000068
- Molecular Weight:171.04 g/mol
- Merck Index:14,1439
- Boiling Pt:179…181 °C
- Melting Pt:–27 °C
- Density:1.420
- Flash Pt:78 °C (172 °F)
- Storage Temperature:Ambient
- EINECS:202-421-7
- UN:3082
- ADR:9,III



